Multi-parameter meters (pH/mV/conductivity/ion), bench, SevenExcellence™ S470/S475
Supplier: Mettler - Toledo
30046252 30046254 30046253 30046255
662-1487EA 3840 EUR
662-1487 662-1485 662-1484 662-1486
Multi-parameter meters (pH/mV/conductivity/ion), bench, SevenExcellence™ S470/S475
Electrochemistry Meters pH and Multi-Parameter Meters
These instruments with two or three channels are modularly expandable with additional measurement parameters at any time.
  • Various conductivity modes, units and temperature correction possibilities offers maximum flexibility
  • Large colour display and intuitive menu guidance in 10 languages
  • Flexible method concept for high reproducibility and security
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
  • Integrated interfaces (USB, RS232 and Ethernet) for data exchange
  • GLP compliant
S470: Professional, yetMore Product Information