pH/mV/°C/ion meter, bench, inoLab® pH/ION 7320
Supplier: WTW
1GA340 1GA330 1GA330P 1GA340P
662-1685EA 3200 EUR
662-1685 662-1683 662-1684 662-1686
pH/mV/°C/ion meter, bench, inoLab® pH/ION 7320
Electrochemistry Meters pH and Multi-Parameter Meters
This two-channel instrument is ideal for precision measurements of pH, ISE or Redox and automatic documentation acc. to GLP/ AQA as required by accredited laboratories. The high resolution measurements can be controlled conveniently and easily via a graphical user interface.
  • Five-point calibration for measuring pH, and up to seven-point for measuring ions
  • USB interface for fast data transfer in .csv format or via the optional integrated printer
  • CMC function for pH and ISE visualises the ideal measuring range and supports correct measuring
  • Graphic display
More Product Information