pH meters, bench, sensION™+ pH3
Supplier: Hach
LPV2011T98002
663-0336EA 1400 EUR
663-0336
pH meters, bench, sensION™+ pH3
Electrochemistry Meters pH and Multi-Parameter Meters
This simple pH meter fulfils all the requirements for routine measurements in industry and education. The all-in-one system with guided menu navigation makes general electrochemistry testing fast and simple. Each system is designed to be used in a wide variety of applications and comes complete with everything you need to start testing.
  • Integrated magnetic stirrer for precise and repeatable measurements in accordance with GLP
  • Labelled calibration flasks minimise the required buffer volume
  • Calibration in 1, 2 or 3 points and mV calibration with Redox
More Product Information