Pocket conductivity/TDS testers, Thermo Scientific™ Eutech™ Elite CTS
Supplier: EUTECH
ELITECTSCUP ELITECTSCUPKIT ELITECTSPIN ELITECTSPINKIT
663-0357EA 160 EUR
663-0357 663-0352 663-0358 663-0351
Pocket conductivity/TDS testers, Thermo Scientific™ Eutech™ Elite CTS
Electrochemistry Meters Conductivity Meters
These rugged, user-friendly testers simultaneously display conductivity/TDS/salinity with temperature. They are available with a pin or cup sensor. The robust cup sensor features an industrial grade SS316 stainless steel sensor element which protects against harsh samples and ensures superior chemical durability. It also allows the user to measure smaller volumes by placing the liquid directly into the cup.
  • Intuitive keypad and large graphic LCD with backlight
  • Auto-hold readings; One-push calibration
  • Easy to replace sensor
  • Leakproof cap with fill-line
More Product Information