Dissolved oxygen meter, bench, inoLab® Oxi 7310
Supplier: WTW
1BA304 1BA300P 1BA301 1BA300
664-0089EA 3970 EUR
664-0089 664-0086 664-0087 664-0085
Dissolved oxygen meter, bench, inoLab® Oxi 7310
Electrochemistry Meters Oxygen Meters
High quality instruments with advanced measuring technology, making measurements more convenient and reliable than ever.
  • AutoRead function ensures repeatable results
  • USB interface for fast data transfer; complete data transfer in .csv format
  • Data output via optional integrated printer
  • Easy to read, graphic display, intuitive user interface and keypad with tactile feedback
The 7310 series is designed for all applications in laboratories where documentation supporting AQA (Analytical Quality Assurance) and GLP (Good Laboratory Practice) is required.