Multi-parameter meters, handheld, U-50 series
Supplier: HORIBA
3200164509 3200164510 3200323680 3200323681 3200323683 3200323686 3200323687 3200164502 3200156563 3200158178 3200164499 3200164501 3200164503 3200164504 3200164505 3200164506 3200164507 3200164508 3200323688 3200164500
665-0336EA 3300 EUR
665-0336 665-0337 665-0338 665-0339 665-0340 665-0341 665-0342 665-0329 665-0393 665-0325 665-0326 665-0328 665-0330 665-0331 665-0332 665-0333 665-0334 665-0335 665-0343 665-0327
Multi-parameter meters, handheld, U-50 series
Electrochemistry Meters pH and Multi-Parameter Meters
These waterproof meters can measure and display up to 11 parameters simultaneously with one single probe. Thanks to their rugged design and their field replaceable sensors they are ideal for outdoor use, for example, when testing ground or surface water. Their compact size and the hand strap enable one-handed operation.
  • Easy to read LCD display with backlight and on-screen icons for battery power, GPS, USB and probe unit connectivity
  • Shock resistant cover, easily cleanable, designed for rough treatment in the field
  • Quick-connect fitting: Cables can easily
More Product Information