Sodium hydroxide 97.0-100.5% Ph. Eur., Macron Fine Chemicals™

Supplier: MACRON AVANTOR BRAND

7066-33EA 30.19 EUR
7066-33 7066-36 7066-38
Sodium hydroxide 97.0-100.5% Ph. Eur., Macron Fine Chemicals™
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR