Butyrometer original gerber
Supplier: Gerber Instruments AG
03.0029
709-0132EA 29 EUR
709-0132
Butyrometer original gerber
Butyrometers
DURAN®, borosilicate glass.
Butyrometers are used to determine the fat content of dairy samples. The product is treated with concentrated sulphuric acid to disrupt the emulsion. The resulting mixture is centrifuged in the butyrometer and the fat content read directly off the scale.