VWR®, Sealing Film for Automation
Supplier: VWR Collection
731-0307EA 778 EUR
731-0307 731-0308 731-0306
VWR®, Sealing Film for Automation
Seals Microplate Seals
Adhesive sealing film or foil rolls constructed on plastic cores, for use with high throughput automated microplate sealers.
  • Continous rolls - no splices
  • Film or foil does not extend beyond edge of plastic core
  • Robust static-free packaging provides protection during shipment and facilitates accurate alignment and adhesion
Sealing film for qPCR, sitting-drop protein crystallisation, HTS and cold storage
Clear polyolefin with ultra-strong, pressure-sensitive silicone adhesive for qPCR, sitting-drop protein crystallisation, HTS, storage and generalMore Product Information