Automation tips for Hamilton® Microlab® Star Line and Nimbus®, Axygen
Supplier: Corning
HT-50-CBK-HTRS HT-1000CBKHTRS HT-10-CBK-HTR HT-10-CBK-HTRS HT-300-CBK-HTR HT-300-CBKHTRS HT-50-CBK-HTR HTF1000CBKHTRS HTF-10-CBKHTRS HTF-300CBKHTRS HTF-50-CBKHTRS HT-10-CBK-STK HT-300-CBK-STK HT-50-CBK-STK HT10CBKHTR5 HT1000CHTRS5 HT10CBKHTRS5 HT50CBKHTR5 HT300CBKHTR5 HT300CBKHTRS5 HT1000CBKHTR5 HT1000CBKHTRS5 HT10CHTR5 HT10CHTRS5 HT50CHTR5 HT300CHTR5 HT300CHTRS5 HT1000CHTR5 HT50CBKHTRS5
731-0423EA 408 EUR
731-0423 731-0416 731-0417 731-0418 731-0420 613-3138 731-0422 731-0425 731-0426 731-0427 731-0428 731-0419 731-0421 731-0424 AXYGHT10CBKHTR5 AXYGHT1000CHTRS5 AXYGHT10CBKHTRS5 AXYGHT50CBKHTR5 AXYGHT300CBKHTR5 AXYGHT300CBKHTRS5 AXYGHT1000CBKHTR5 AXYGHT1000CBKHTRS5 AXYGHT10CHTR5 AXYGHT10CHTRS5 AXYGHT50CHTR5 AXYGHT300CHTR5 AXYGHT300CHTRS5 AXYGHT1000CHTR5 AXYGHT50CBKHTRS5
Automation tips for Hamilton® Microlab® Star Line and Nimbus®, Axygen
Pipette Tips
Axygen brand automation tips are compatible with Hamilton® Microlab® Star and Nimbus® robotic systems.
  • Conductive black or clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
Tips are available in 4 sizes (10, 50, 300, and 1000 µl) and are packaged in either traditional hanging tip racks ('frame' style) or deck-seating nested tip racks. As with all Axygen robotic tips, they are manufactured to stringent specifications under strict process control and assembled via automation to ensure consistent performance and quality.