PCR plates, 96-well, low profile, half skirt, Axygen®
Supplier: Corning
PCR-96-LP-AB-C PCR-96-LP-AB-W PCR-96LPABCPBC
732-1015EA 224 EUR
732-1015 732-1017 AXYGPCR-96LPABCPBC
PCR plates, 96-well, low profile, half skirt, Axygen®
Microplates PCR Plates
Axygen® 96 well, PP, low profile PCR microplates are designed to fit ABI Real-Time PCR thermal cyclers and sequencers.
  • Optimised for Fast PCR: Low profile design results in faster and more efficient PCR reactions
  • Reduced volume minimises potential evaporation
  • Ultra-thin and consistent wall thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified RNase, DNase and endotoxin-safe for highest sample integrity
  • Can be sealed with 8/12 strip caps, silicone AxyMat™ or sealing film
  • AxyMat™ sealing mats autoclavable
More Product Information