Corning® Treated Culture Dishes
Supplier: Corning
430165 430166 430167 430196 430293 430599 431110 3261 3295 4615
734-1698EA 335 EUR
734-1698 734-1699 734-1815 734-1703 734-1705 734-1711 734-1727 734-0884 734-4056 CORN4615
Corning® Treated Culture Dishes
Culture Dishes Cell Culture Dishes
Optically clear PS. Available with a choice of treated surface. Corning® CellBIND® surface increases surface wetability for more even and consistent cell attachment. Ultra-Low Attachment dishes feature a covalently bound hydrogel layer that minimises cell attachment, protein absorption and cellular activation.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilised by gamma radiation
  • Certified non-pyrogenic
These disposable petri-style dishes are manufactured from optically clear virgin polystyrene. DishesMore Product Information