Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
430052 430053 430055 430766 430790 430791 431470
734-1812EA 231 EUR
734-1812 734-1813 734-1814 734-1862 734-1866 734-1867 525-0477
Centrifuge tubes, 15 ml, Corning®
Tubes Centrifuge Tubes
Corning® 15 ml clear centrifuge tubes are provided without caps for ease of use in automated or other applications where non pre-attached caps are desired.
  • Black printed graduations and a large white marking spot
  • Maximum RCF is 12000×g
  • Sterile, certified nonpyrogenic and DNase-/RNase-free
The CentriStar™ cap has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping. This leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when usedMore Product Information