Serological pipettes, Nunc™
Supplier: Thermo Scientific
170353 170354 170356 170358 170364 170365 170366 170367 170368 170369 170375 170376
612-4220EA 258 EUR
612-4220 612-4221 612-4223 612-4225 734-2612 734-2613 734-2614 734-2615 734-2616 734-2617 734-2622 734-2623
Serological pipettes, Nunc™
Pipettes Serological Pipettes
PS, sterile.
  • Coloured top, plugged, coloured packaging for easy identification
  • Convenient, black, extra graduations to full pipette volume
  • Non pyrogenic
These accurate, disposable, plastic pipettes are supplied plugged and sterilised.