Supelco® TraceCERT® Organic Reference Standard, Dicofol
Supplier: SUPELCO
74046-50MG
74046-50MGEA 205 EUR
74046-50MG
Supelco® TraceCERT® Organic Reference Standard, Dicofol
Standards Organic Standards
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352 + P312