Avantor® Seradigm Australia Origin Foetal Bovine Serum (FBS)
Supplier: VWR Seradigm
76294-120EA 189 EUR
76294-120 76294-180 76294-178 76294-122 76294-124 76294-182
Avantor® Seradigm Australia Origin Foetal Bovine Serum (FBS)
Sera (Serum)
Australia Origin FBS is widely considered the gold standard for safety and regulatory compliance by cell culturists around the world. Avantor Seradigm Australia Origin FBS combines outstanding performance and quality with ease of regulatory access globally, suitable for a wide variety of applications.
  • 100% Australia origin
  • Australian disease-free status
  • Preservation of native, growth-promoting factors
  • Single-use sterile filtration technology
  • Proven performance and consistency
Avantor Seradigm is supplier of animal sera that provides the cell cultureMore Product Information