Phosphatase inhibitor cocktails, PhosphataseArrest™
Supplier: G Biosciences
786-450 786-451 786-452 786-647 786-784 786-783 786-782
786-450EA 578 EUR
786-450 786-451 786-452 786-647 786-784 786-783 786-782
Phosphatase inhibitor cocktails, PhosphataseArrest™
Cell Culture Supplements and Additives
The PhosphataseArrest™ phosphatase inhibitor cocktails are ready to use 100X solutions that are simply added to either extraction buffers or samples.
  • Single 100X solution
  • Ready to use
  • Compatible with most phosphatase assays
  • No resuspension required
PhosphataseArrest™ I: A broad spectrum phosphatase inhibitor cocktail consisting of five phosphatase inhibitors that target serine/threonine specific, tyrosine specific and dual specificity phosphatases. PhosphataseArrest™ I is a stablised solution of sodium fluoride, sodium orthovanadate, sodium pyrophosphate, β-glycerophosphate & sodium molybdate.
PhosphataseArrest™ II: A phosphatase inhibitor cocktail consisting of five phosphatase inhibitors that target acid, alkaline and tyrosine phosphatases. PhosphataseArrest™ II contains optimised concentrations of sodium fluoride, sodium tartrate, sodium orthovanadate, imidazole and sodium molybdate.
PhosphataseArrest™ III: A phosphatase inhibitor cocktail consisting of three phosphatase inhibitors that target alkaline and serine/threonine phosphatases. PhosphataseArrest™ III is a stable, convenient 100X solution of cantharidin, bromotetramisole, and microcystin LR.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more