Anthrone ≥97.0% for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Warning


MQ200

8.01461.0010
8.01461.0010EA 53.2 EUR
8.01461.0010
Anthrone ≥97.0% for synthesis, Sigma-Aldrich®
Anthrone
Formula: C₁₄H₁₀O
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001187
CAS Number: 90-44-8
EINECS: 201-994-0

Order Now

Specification Test Results

CAS 90-44-8
Synonyms 9,10-Dihydro-9-oxoanthracene
Hill Formula C₁₄H₁₀O
Molar Mass 194.23 g/mol
Melting Point 151 - 156 °C
Bulk density 450 kg/m³
Solubility 4.6 mg/l (20 °C)
Assay (GC, area%) ≥ 97,0 Area %
Identity (IR) conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR