Mesitylene for synthesis

Supplier: MERCK MILLIPORE
Danger

8.05890.1000 8.05890.9021 8.05890.6165
8.05890.1000EA 83.6 EUR
8.05890.1000 8.05890.9021 8.05890.6165
Mesitylene for synthesis
Mesitylene
Formula: C₉H₁₂
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0,8637 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00008538
CAS Number: 108-67-8
EINECS: 203-604-4
UN: 2325
ADR: 3,III
Merck Index: 13,05934

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR