Methylammonium chloride, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ200

8.06020.0005 8.06020.0250 8.06020.1000 8.06020.9025 8.06020.9032
8.06020.0005EA 30 EUR
8.06020.0005 8.06020.0250 8.06020.1000 8.06020.9025 8.06020.9032
Methylammonium chloride, Sigma-Aldrich®
Methylammonium chloride
Formula: CH₆ClN
MW: 67.52 g/mol
MDL Number: MFCD00012849
CAS Number: 593-51-1
EINECS: 209-795-0

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 228 - 233 °C
Molar Mass 67.52 g/mol
Bulk Density 1100 kg/m3
Boiling Point 225 - 230 °C (20 hPa)
pH 5 - 7 (10 g/l, H2O, 20 °C)
Hill Formula CH6ClN
Chemical Formula CH3NH3Cl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR