2-(2-Naphthoxy)ethanol for synthesis

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ200

8.14891.0005
8.14891.0005EA 217 EUR
8.14891.0005
2-(2-Naphthoxy)ethanol for synthesis
2-(2-Naphthoxy)ethanol
Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
MDL Number: MFCD00016809
CAS Number: 93-20-9
EINECS: 202-228-8

Order Now

Specification Test Results

Melting Point 75 °C
Molar Mass 188.23 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR