L(+)-Valine for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ200

8.16006.0025 8.16006.0100
8.16006.0025EA 41.1 EUR
8.16006.0025 8.16006.0100
L(+)-Valine for synthesis, Sigma-Aldrich®
L(+)-Valine
Formula: C₅H₁₁NO₂
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 86 °C
Density: 1,284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064220
CAS Number: 72-18-4
EINECS: 200-773-6
Merck Index: 13,09458

Order Now

Specification Test Results

Solubility 85 g/l (20 °C)
Melting Point 315 °C
Molar Mass 117.15 g/mol
Bulk Density 400 kg/m3
Density 1.23 g/cm3 (20 °C)
pH 5.5 - 6.5 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR