Hexamethylenetetramine ≥99.0% for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ200

8.18712.0100 8.18712.1000 8.18712.5000 8.18712.9050
8.18712.0100EA 21.5 EUR
8.18712.0100 8.18712.1000 8.18712.5000 8.18712.9050
Hexamethylenetetramine ≥99.0% for synthesis, Sigma-Aldrich®
Methenamine
Formula: C₆H₁₂N₄
MW: 140.19 g/mol
Melting Pt: 280
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 250
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
EINECS: 202-905-8
UN: 1328
ADR: 4.1 III
Merck Index: 13,05994
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ecbb5df5394322c2150525ce6e9f48b5aadde7&vid=a3ecbb5df5394322c2150525ce6e9f48b5aadde7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HClO₄) ≥99.0%
Identity (IR) Passes Test
Bulk Density 600 kg/m3
Vapor Pressure (20 °C)
Flash Point 250 °C
Density 1.33 g/cm3 (20 °C)
pH 7.0 - 9.0 (100 g/l, H2O, 20 °C)
Ignition Point 390 °C
Hazard Class 4.1
Hazard Class Description Flammable Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR