Sodium butyrate for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ200

8.20236.0250 8.20236.1000
8.20236.0250EA 130 EUR
8.20236.0250 8.20236.1000
Sodium butyrate for synthesis, Sigma-Aldrich®
Sodium butyrate
Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: 244
Density: 1.324 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 244 °C
Molar Mass 110.09 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR