1-Dodecylpyridinium chloride, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ200

8.20546.0005 8.20546.0250 8.20546.1000 8.20546.9025
8.20546.0005EA 43.1 EUR
8.20546.0005 8.20546.0250 8.20546.1000 8.20546.9025
1-Dodecylpyridinium chloride, Sigma-Aldrich®
1-Dodecylpyridinium chloride
Formula: C₁₇H₃₀ClN
MW: 283.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011987
CAS Number: 104-74-5
EINECS: 203-232-2
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Solubility 600 g/l (20°C)
Melting Point 87 - 89 °C
Flash Point 160 °C
Density 1.04 g/cm3 (20 °C)
pH 4 (10 g/l, H2O, 20 °C)
Ignition Point 270 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR