5-(Hydroxymethyl)-2-furaldehyde for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ200

8.20678.0001 8.20678.0005 8.20678.0200 8.20678.8100
8.20678.0001EA 47.6 EUR
8.20678.0001 8.20678.0005 8.20678.0200 8.20678.8100
5-(Hydroxymethyl)-2-furaldehyde for synthesis, Sigma-Aldrich®
5-(Hydroxymethyl)-2-furaldehyde
Formula: C₆H₆O₃
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 27…30 °C
Density: 1,24 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003234
CAS Number: 67-47-0
EINECS: 200-654-9
Merck Index: 13,04857

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 27 - 30 °C
Molar Mass 126.11 g/mol
Boiling Point 154 - 155 °C (16 hPa)
Flash Point 79 °C
Refractive Index 1.5627 (20 °C, 589 nm)
Synonyms 5-(Hydroxymethyl)furfurol
Hill Formula C6H6O3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR