N-phenylanthranilic acid for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE


M-Clarity™ quality level = MQ200

8.20979.0100
8.20979.0100EA 70.8 EUR
8.20979.0100
N-phenylanthranilic acid for synthesis, Sigma-Aldrich®
N-phenylanthranilic acid
Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Boiling Pt: 375
Melting Pt: 182…185 °C
MDL Number: MFCD00002421
CAS Number: 91-40-7
EINECS: 202-066-8
Merck Index: 12,07427

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 182 - 185 °C
Molar Mass 213.23 g/mol
Bulk Density 200 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR