4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ200

8.40130.0025 8.40130.0100
8.40130.0025EA 63.9 EUR
8.40130.0025 8.40130.0100
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) for synthesis, Sigma-Aldrich®
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
Formula: C₉H₁₈NO₂
Storage Temperature: Ambient
MDL Number: MFCD00006478
CAS Number: 2226-96-2
EINECS: 218-760-9

Order Now

Specification Test Results

Solubility 1670 g/l (20 °C)
Melting Point 67 - 71 °C
Molar Mass 172.25 g/mol
Boiling Point (1013 hPa) not applicable,(decomposition)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 146 °C DIN 51758
pH 8.2 (20 g/l, H2O, 20 °C)
Ignition Point 260 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR