3-Mercapto-1,2-propanediol for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ200

8.41705.0050 8.41705.0250 8.41705.1000 8.41705.3000 8.41705.0010
8.41705.0050EA 79.4 EUR
8.41705.0050 8.41705.0250 8.41705.1000 8.41705.3000 8.41705.0010
3-Mercapto-1,2-propanediol for synthesis, Sigma-Aldrich®
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
Boiling Pt: 118 °C (1013 hPa)
Density: 1,247 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
UN: 2810
ADR: 6.1,III
Merck Index: 12,09471

Order Now

Specification Test Results

Molar Mass 108.15 g/mol
Boiling Point 118 °C (6.7 hPa)
Flash Point 110 °C
Density 1.247 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR