Ethyl-3-methoxypropionate for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Warning


M-Clarity™ quality level = MQ200

8.43946.1000
8.43946.1000EA 69.6 EUR
8.43946.1000
Ethyl-3-methoxypropionate for synthesis, Sigma-Aldrich®
Ethyl 3-methoxypropionate
Formula: C₆H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00143172
CAS Number: 10606-42-5
EINECS: 212-112-9
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR