Storage solution for ROSS electrodes
Supplier: THERMO ORION
810001
810001.EA 154 EUR
810001.
Storage solution for ROSS electrodes
Electrolytes Electrode Storage Solutions
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH electrode storage solutions.
  • Storage solution ensures that the pH electrode works properly after prolonged storage, overnight storage and between samples