VWR® PolarSafe Dots, Cryo-Labels
Supplier: VWR Collection
817-0153EA 63 EUR
817-0153 817-0155 817-0154 817-0156
VWR® PolarSafe Dots, Cryo-Labels
Labels
PolarSafe Dots are made from chemically inert polyester with a unique adhesive formulated to adhere to laboratory glass and plastic ware. These dots will not only withstand conventional freezing (−80 °C) but can also be used for most cryogenic storage applications (vapour phase nitrogen storage at −196 °C). No need for separate labelling solutions for your cryo-preservation units and your ultra low freezers. The labels can even be used in incubators. PolarSafe Dots do not require self lamination to be effective. The sleeker labels mean your tubes will still fit most storage systems withoutMore Product Information