Cleanroom tapes, MicroSeal™ CSLB™
Supplier: MICRONOVA
CSLB-2WHIR CSLB-236WH CSLB-34WH CSLB-3WH CSLB-3WHIR CSLB-1WHIR CSLB-12WHIR CSLB-34WHIR CSLB-12WH CSLB-1WH
819-0025EA 1330 EUR
819-0025 819-0157 819-0096 819-0098 819-0105 819-0095 819-0155 819-0158 819-0154 819-0156
Cleanroom tapes, MicroSeal™ CSLB™
Tapes
These cleanroom tapes are made of PE with pressure-sensitive acrylic adhesive.
  • Pre-perforated
  • Perforations at 7,6 cm (3") intervals allow an easy tear at optimum lengths
  • Low-density, low-outgassing polyethylene film will remove cleanly, leaving no adhesive residue
  • Can be written on with fibre tip and permanent markers
  • Wound on a 3" plastic core with no liner, meaning less packaging and waste to discard
  • Available from 0.5 to 3" widths; smaller widths are ideal for rodac plate sealing during environmental monitoring
  • Suitable for food processing and
More Product Information