Washi masking tape 244, Scotch®
Supplier: 3M
244-18MM 244-24MM 244-36MM 244-48MM
819-0118EA 15.5 EUR
819-0118 819-0119 819-0120 819-0121
Washi masking tape 244, Scotch®
Tapes
This is provided with a yellow smooth paper backing and a 0,085 mm thick synthetic adhesive. This tape is particularly resistant to UV, and is waterproof.
  • Excellent UV resistance for outdoor use (max. six weeks)
  • Clean removal, even after six weeks from most surfaces
  • Ultra-flat paint edges and extremely thin backing material
  • Resists paint bleed
  • Easily removed after heating, without leaving residues
  • Temperature-resistant up to 100 °C for 30 minutes
This tape can be used for a wide range of masking applications, especially for where UV resistance andMore Product Information