Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Danger

8254.1000EA 72.75 EUR
8254.1000 8254.1000S 8254.5000 8254.9025
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Diethyl ether
Formula: (CH₃CH₂)₂O
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0,7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=e25ba31ba5c9afb69382c3ca3d66e401980e4bba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay (by GC) min. 99.5 %
Alcohol (C2H5OH) max. 0.01 %
Carbonyl Compounds (as HCHO) max. 0.001 %
Color (APHA) max. 10
Density (g/ml) at 20°C 0.712 - 0.714
Foreign Odor passes test
Peroxide (as H2O2) max. 3 ppm
Residue after Evaporation max. 0.001 %
Substances Darkened by H2SO4 passes test
Titrable Acid (meq/g) max. 0.0002
Water (H2O) max. 0.01 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR