Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

83684.230EA 164 EUR
83684.230 83684.290 83684.269
Pyridine, anhydrous (max. 0.003% H₂O) ≥99.8%
Pyridine

250 and 500 ml bottles with a septum cap featuring six separate re-sealable puncture points

Formula: C₅H₅N
MW: 79.1 g/mol
Boiling Pt: 115
Melting Pt: –44 °C
Density: 0.983 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3 II
Merck Index: 13,08060
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=5dc7f822262726db0427fe8ab02ba2d8cf31c801&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8 %
Evaporation residue Max. 3 ppm
Water Max. 30 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR