2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC

Supplier: VWR Chemicals
Danger


Filtered through a 0.2 µm filter, packaged under nitrogen.

84531.550EA 0 EUR
84531.550 84531.460 84531.950
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR