Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)

Supplier: VWR Chemicals

85830.290EA 38 EUR
85830.290 85830.360 85830.410 85830.460 85830.550
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Ethanol Eurodenatured
Formula: H₃CCH₂OH
MW: 46.07 g/mol
CAS Number: 64-17-5
UN: 1170
ADR: 3 II

Order Now

Specification Test Results

Assay (V/V) min. 97%
Assay (GC) min. 99%
Denatonium benzoate (ppm) 12.3 - 14.7 ppm
Methyl ethyl ketone (V/V) 0.97 - 1.13 %
2-Propanol (V/V) 0.97 - 1.13%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR