1,2-Dichlorobenzene ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

85995.290EA 68 EUR
85995.290 85995.320 85995.180
1,2-Dichlorobenzene ≥99%, GPR RECTAPUR®
1,2-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 180.5
Melting Pt: -17
Flash Pt: 66
MDL Number: MFCD00000535
CAS Number: 95-50-1
UN: 1591
ADR: 6.1 III
Merck Index: 13,03081

Order Now

Specification Test Results

Assay Min. 99%
Density (20/4) 1.305 - 1.307


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR