Miniature inert valves, HV series
Supplier: HAMILTON BONADUZ
86725 86726 86727 86728 86729 86730 86731 86732 86735 86736 86738 86788 86779 86777 86901 86903 86915 86778 86781 86775 86780 86918 86785 86786 86787 86902 86905 86919 86907 86910 86911 86912 86913 86916 86904
86725.EA 158 EUR
86725. 86726. 86727. 86728. 86729. 86730. 86731. 86732. 86735. 86736. 86738. 86788. R86779 R86777 R86901 R86903 R86915 R86778 R86781 R86775 R86780 R86918 R86785 R86786 R86787 R86902 R86905 R86919 R86907 R86910 R86911 R86912 R86913 R86916 R86904
Miniature inert valves, HV series
Valves Micro Valves
HV plug valves provide an economical and flexible method of custom-building a fluid flow control system, from a simple on/off valve to a more complex multi-valve arrangement, using a combination of loop and distribution valves.
  • Wetted paths are chemically inert PTFE and Kel-F® materials
  • Valve position is controlled manually via a handle attached to the valve stem
  • Valve can be chosen by selecting the proper flow path
The small valve body with its chemically inert flow path is ideally suited for low pressure applications. All HV Plug Valves allow you toMore Product Information