, pellets, Ø 1 - 2 mm (0.04 - 0.08 in)

Supplier: ALFA AESAR

87957.A1 87957.A7
87957.A1EA 138 EUR
87957.A1 87957.A7
, pellets, Ø 1 - 2 mm (0.04 - 0.08 in)
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR