Digital timer
Supplier: KARTELL
902
902.EA 112 EUR
902.
Digital timer
Timers Digital Timers
Electronic timer, battery operated.
  • Four digits display up to 19 hours 59 minutes
  • Alarm sounds at the end of the set time
  • Free-standing, clip or magnetic fixings