2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIALS
Danger


Management of Change (MOC) category = R

9059-08EA 0 EUR
9059-08 9059-09 9059-10
2-Propanol ≥99.5%, CMOS for microelectronic, J.T.Baker®
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=77f11bcf690db67b8cf7c1dbbc5d3a4d2586aeed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Microelectronic Use = 99.5 %
Assay (CH₃CHOHCH₃) ≥ 99.5 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 4 ppm
Solubility in H₂O Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤ 0.05 %
Acidity (µeq/g) ≤ 0.2
Alkalinity (µeq/g) ≤ 0.1
Chloride (Cl) ≤ 0.2 ppm
Phosphate (PO₄) ≤ 0.3 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 10.0 ppb
Trace Impurities - Barium (Ba) ≤ 20.0 ppb
Trace Impurities - Beryllium (Be) ≤ 100.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 100.0 ppb
Trace Impurities - Boron (B) ≤ 10.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 20.0 ppb
Trace Impurities - Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Chromium (Cr) ≤ 20.0 ppb
Trace Impurities - Cobalt (Co) ≤ 20 ppb
Trace Impurities - Copper (Cu) ≤ 10.0 ppb
Trace Impurities - Gallium (Ga) ≤ 50 ppb
Trace Impurities - Germanium (Ge) ≤ 50.0 ppb
Trace Impurities - Gold (Au) ≤ 20 ppb
Heavy Metals (as Pb) ≤ 200.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lead (Pb) ≤ 20.0 ppb
Trace Impurities - Lithium (Li) ≤ 50.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 20 ppb
Trace Impurities - Manganese (Mn) ≤ 15.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 100.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 100.0 ppb
Trace Impurities - Potassium (K) ≤ 100 ppb
Trace Impurities - Silicon (Si) ≤ 50 ppb
Trace Impurities - Silver (Ag) ≤ 20.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 20.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 100.0 ppb
Trace Impurities - Thallium (Tl) ≤ 10.0 ppb
Trace Impurities - Tin (Sn) ≤ 100 ppb
Trace Impurities - Titanium (Ti) ≤ 20 ppb
Trace Impurities - Vanadium (V) ≤ 100.0 ppb
Trace Impurities - Zinc (Zn) ≤ 50 ppb
Trace Impurities - Zirconium (Zr) ≤ 100.0 ppb
Particle Count at point of fill - 0.5 µm and greater (Rion KS42AF) ≤ 150 par/ml
Particle Count at point of fill - 1.0 µm and greater (Rion KS42AF) ≤ 25 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR