VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Supplier: VWR Chemicals
91021.050EA 205 EUR
91021.050 91024.050 91025.050 91026.050 91027.050 91028.050 91029.050 91031.050 91029.010 91031.010 91021.010 91024.010 91026.001 91027.010 91028.010 91029.001 91072.050 91073.010 91073.050 91074.010 91074.050 91076.010 91076.050 91078.010 91078.050 91080.010 91080.050 91082.010 91082.050 91083.002 91083.010 91083.050 91087.002 91087.010 91087.050 91088.001 91088.010 91088.050 91089.001 91090.001 91090.010 91090.050 91091.001 91091.002 91091.010 91072.010 92156.001 91257.250 91255.001 91251.010 91254.001 91249.100 91252.100 91250.001 91253.001
VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Standards Chromatography Standards
Avantor offers an extensive range of FAME standards as Certified Reference Materials under the well-known PESTINORM® brand.
  • All products are CRM acc ISO/IEC 17025 and ISO 17034
  • All products are NEAT
  • 24 month shelf life
Esterification of fatty acids is an important tool to determine content and characterise fats and oils in food, especially for measurement by GC methods. The simple derivatisation to create the methyl ester analogue is also used widely as a preliminary technique for assessing the quality and purity of biofuels. VWR® offers a comprehensive range of fatty acid methyl esters (FAME) Certified Reference Materials for calibration of GC instruments, for validation of analytical methods, preparation of 'working reference samples' or for detection limit and linearity studies.
Certifications: ISO 17025 / 17034