VWR®, PE Pure 10, 23 components in 2% HNO₃
91195.180EA 359 EUR
91195.180
VWR®, PE Pure 10, 23 components in 2% HNO₃
Standards Cationic Standards Multi-Element Standards
23 components: As 50 ug/l, B 100 ug/l, Ba 50 ug/l, Be 20 ug/l, Bi 10 ug/l, Ca 35000 ug/l, Cd 20 ug/l, Co 25 ug/l, Cr 20 ug/l, Cu 20 ug/l, Fe 100 ug/l, K 3000 ug/l, Mg 15000 ug/l, Mn 30 ug/l, Mo 100 ug/l, Na 8000 ug/l, Ni 50 ug/l, Pb 25 ug/l, Se 10 ug/l, Sr 100 ug/l, Tl 10 ug/l, V 50 ug/l, Zn 50 ug/l in 2% HNO₃
Certifications: ISO 17025/17034