VWR®, Standard Solution, 7 components: 10 ug/ml each of 6Li, Sc, Ge, Y, in, Tb, Bi in 5% HNO₃
91208.180EA 228 EUR
91208.180
VWR®, Standard Solution, 7 components: 10 ug/ml each of 6Li, Sc, Ge, Y, in, Tb, Bi in 5% HNO₃
Standards Cationic Standards Multi-Element Standards
Certifications: ISO 17025/17034