VWR®, Combined Seven Anion Standard I , 7 components in H₂O
91292.180EA 212 EUR
91292.180 91292.154
VWR®, Combined Seven Anion Standard I , 7 components in H₂O
Standards Anionic Standards Multi-Element Anioinic Standards
7 components: Fluorides (F-) 20 mg/l, Chlorides (Cl-) 30 mg/l, Nitrites (NO2-) 100 mg/l, Bromides (Br-) 100 mg/l, Nitrates (NO3-) 100 mg/l, Phosphates (PO43-) 150 mg/l, Sulphates (SO42-) 150 mg/l in H₂O
Certifications: ISO 17025/17034