Supelco® TraceCERT® Organic Reference Standard, ʟ-Tyrosine

Supplier: SUPELCO

91515-100MG
91515-100MGEA 88.5 EUR
91515-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Tyrosine
Standards Organic Standards

  • GHS signal: GHS07


Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.

https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b43b3f5c113f68a1bb8ea07bd9476955c541c96b&vid=b43b3f5c113f68a1bb8ea07bd9476955c541c96b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid; 3-(4-Hydroxyphenyl)-ʟ-alanine
Environmentally Preferable
CAS No. 60-18-4
MDL No. MFCD00002606
Formula 4-(HO)C₆H₄CH₂CH(NH₂)CO₂H
Molecular weight 181,19
Melting point >300 °C (dec.) (lit.)
Shipping conditions STD
Storage conditions 15...25 °C
M-Clarity quality level MQ300

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR