Acrylamide : Bisacrylamide (29:1) 40% in aqueous solution

Supplier: APPLICHEM LIFESCIENCE
Danger

A0385.0500 A0385.1000
A0385.0500EA 102 EUR
A0385.0500 A0385.1000
Acrylamide : Bisacrylamide (29:1) 40% in aqueous solution
Acrylamide-Bis (32:1)
Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR