Lecithin from egg yolk

Supplier: APPLICHEM LIFESCIENCE

A0893.0100 A0893.0500
A0893.0100EA 216 EUR
A0893.0100 A0893.0500
Lecithin from egg yolk
Lecithin from egg yolk
MDL Number: MFCD00131449
CAS Number: 93685-90-6
EINECS: 297-639-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR